Order Entry
Austria
ContactUsLinkComponent
5-Tetramethylrhodamine-dUTP, 1 mM Solution
5-Tetramethylrhodamine-dUTP, 1 mM Solution
Catalog # 40001.
Supplier:  Biotium
CAS Number:  
5-Tetramethylrhodamine-dUTP, 1 mM Solution
Catalog # 40001.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 µl
  • Description:
    5-Tetramethylrhodamine-dUTP, 1mM Solution
  • Storage conditions:
    –20°

Specifications

About this item

5-TAMRA-dUTP (5-Tetramethylrhodamine-dUTP) can be enzymatically incorporated into DNA to make fluorescently labeled probes, and has absorption/emission at 553/577 nm. Note: 5-TAMRA-dUTP may not be compatible with PCR labeling of DNA probes. Supplied as a 1 mM solution in Tris-HCl buffer, pH 7.5.