Order Entry
Austria
ContactUsLinkComponent
Isohexane (mixture of isomers)
Isohexane (mixture of isomers)
Catalog # ROTHT176.1
Supplier:  Roth Carl
CAS Number:  
Isohexane (mixture of isomers)
Catalog # ROTHT176.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

About this item

 :
 :86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–154 °C
Density:0.653 g/cm³ (25 °C)
Flash Pt:–33 °C
Storage Temperature:Ambient
EINECS:295-570-2
UN:1208
ADR:3,II
REACH:01-2119484651-34
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:92112-69-1
 :73513-42-5
Formula:C₆H₁₄
MDL Number:MFCD00009406