Order Entry
Austria
ContactUsLinkComponent
Isohexane (mixture of isomers), SupraSolv® for gas chromatography, Supelco®
Isohexane (mixture of isomers), SupraSolv® for gas chromatography, Supelco®
Catalog # 1.04340.2500
Supplier:  Merck
CAS Number:  
Isohexane (mixture of isomers), SupraSolv® for gas chromatography, Supelco®
Catalog # 1.04340.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    0.02 g/l (20 °C)
  • Melting Point:
    86.18 g/mol
  • Molar Mass:
    0.66 g/cm3 (20 °C)
  • Boiling Point:
    53 - 63 °C (1013 hPa)
  • Vapor Pressure:
    160 - 190 hPa (20 °C)
  • Flash Point:
    -21 °C
  • Explosion Limit:
    1.0 - 7.4 %(V)
  • pH:
    (H2O) no data available
  • Ignition Point:
    260 °C
  • Viscosity kinematic:
    0.5 mm2/s (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–154 °C
Density:0.653 g/cm³ (25 °C)
Flash Pt:–33 °C
Storage Temperature:Ambient
EINECS:295-570-2
UN:1208
ADR:3,II
 :M-Clarity™ quality level = MQ100
REACH:01-2119484651-34
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:92112-69-1
 :73513-42-5
Formula:C₆H₁₄
MDL Number:MFCD00009406