Isohexane (mixture of isomers), EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:92112-69-1
:73513-42-5
Formula:C₆H₁₄
MDL Number:MFCD00009406
:86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–154 °C
Density:0.653 g/cm³ (25 °C)
Flash Pt:–33 °C
Storage Temperature:Ambient
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ300
EINECS:295-570-2
REACH:01-2119484651-34
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:0.02 g/l (20 °C)
- Molar Mass:86.18 g/mol
- Boiling Point:53 - 63 °C (1013 hPa)
- Vapor Pressure:160 - 190 hPa (20 °C)
- Flash Point:-21 °C
- Explosion Limit:1.0 - 7.4 %(V)
- Density:0.66 g/cm3 (20 °C)
- Ignition Point:260 °C
- Viscosity kinematic:0.5 mm2/s (20 °C)
Specification Test Results
Frequently Bought Together