Order Entry
Austria
ContactUsLinkComponent
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Catalog # 34493-2.5L
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Catalog # 34493-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay of isomers (GC):
    Min. 85 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • n-Hexane (GC):
    Max. 4.9 %
  • Explosion Limit:
    7.5 %
  • Autoignition Temperature:
    437 °F

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:48…65 °C (1.013 hPa)
Melting Pt:–154 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–26 °C
Storage Temperature:Ambient
EINECS:295-570-2
UN:1208
ADR:3,II
REACH:01-2119484651-34
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311