Order Entry
Austria
ContactUsLinkComponent
VWR® Bouin's solution
VWR® Bouin's solution
Catalog # 7000.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Bouin's solution
Catalog # 7000.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Bouin's solution

Specifications

About this item

VWR® Bouin tissue fixative solution.

Specification
Composition: Passes test