Formaldehyde 7,5%, TECHNICAL, buffered (pH 7.0 ±0.2)
Supplier: VWR Chemicals
About this item
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
2 Options Available Below
Product Details & Documents
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
Specifications for Product Family
Specification Test Results
- Assay (Formaldehyde):7 - 9 %
- pH (20°C):6.8 - 7.2



