Universal pH indicator in ethanol, Technical (pH 3 - 11)
Catalog # 34352.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Identification:Passes test
- Red:pH 3
- Red-orange:pH 4
- Orange :pH 5
- Yellow :pH 6
- Yellow-green:pH 7
- Green :pH 8
- Green-blue:pH 9
- Violet :pH 10
- Violet-red:pH 11
Specification Test Results
About this item
:
Flash Pt:20 °C
ADR:3,II