
Meyer's reagent for blood research
Catalog # 31072.268
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Boiling Pt:>100 °C (1013 hPa)
Density:1.1 g/cm³ (20 °C)
UN:1760
ADR:8,II
:GHS07