Order Entry
Austria
ContactUsLinkComponent
Schiff reagent for analysis of aldehydes
Schiff reagent for analysis of aldehydes
Catalog # 30969.261
Supplier:  VWR Chemicals
CAS Number:  
undefined
Schiff reagent for analysis of aldehydes
Catalog # 30969.261
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Sensitivity:
    Passes test
  • Density (20/4):
    1.005 - 1.015

Specification Test Results

About this item

 :
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
MDL Number:MFCD00081985