

o-Tolidine 0.1% 1.6 M hydrochloric solution
Catalog # 28672.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:212.29 g/mol
EINECS:204-358-0
UN:1760
ADR:8,III
:GHS08
:GHS07
CAS Number:119-93-7
Formula:[-C₆H₃(CH₃)-₄-NH₂]₂
MDL Number:MFCD00014773