Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lecithin
Lecithin (from soy bean), powder, GPR RECTAPUR®
Lecithin (from soy bean), powder, GPR RECTAPUR®
24967.183
:
VWR Chemicals
:
Lecithin (from soy bean), powder, GPR RECTAPUR®
24967.183
:
VWR Chemicals
:
Pack type:
Plastic bottle for solids
Pk:
100 g
Seq:
3401
Insolubility in acetone:
Min. 62 %
Peroxide value:
Max. 5
:
:
310-129-7
:
8030-76-0
:
MFCD00147406