Order Entry
Austria
ContactUsLinkComponent
Lecithin from egg yolk, GPR RECTAPUR®
Lecithin from egg yolk, GPR RECTAPUR®
Catalog # 24966.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lecithin from egg yolk, GPR RECTAPUR®
Catalog # 24966.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Loss on drying (100°C):
    Max. 6 %
  • N (Nitrogen):
    1.3 - 1.9 %
  • P (Phosphorus) (on dry substance):
    Min. 3 %
  • Solubility in ethanol 96 %:
    Min. 90 %

Specification Test Results

About this item

 :
EINECS:297-639-2
CAS Number:93685-90-6
MDL Number:MFCD00131449