ContactUsLinkComponent
Silicone oil, Technical Grade for oil baths, VWR®
Silicone oil, Technical Grade for oil baths, VWR®
  24610.363
 :  
undefined
Silicone oil, Technical Grade for oil baths, VWR®
  24610.363
 :  
  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190
 :
 :0,96 g/cm³ (20 °C)
 :321 °C (closed cup)
 :Ambient
 :68083-14-7
 :63148-62-9
 :MFCD00132673