Order Entry
Austria
ContactUsLinkComponent
 
Ethanol denatured (industrial methylated spirit) ≥99%, GPR RECTAPUR® (denatured with methanol)
 
undefined
Ethanol denatured (industrial methylated spirit) ≥99%, GPR RECTAPUR® (denatured with methanol)
 

 

  • Environmentally Preferable:

 

 

  • ALCH:
    99.1 - 99.5 %
  • Density (20/4):
    0.792 - 0.794 g/cm3
  • Colour number (Hazen):
    Max. 20.0 APHA

 

Frequently Bought Together