VWR®, Casein
22544.292
:
- Pack type:Plastic bottle
- Pk:1 kg
- CAS No.:9000-71-9
- Storage conditions:Ambient
- Protein/peptide name:Casein
- Grade:Technical Grade
Proteins (on anhydrous product): ≥ 90,0 %
Free acidity: Max. 1,0%
Fats: Max. 2,5%
Ignition residue (SO₄): Max. 2,75%
Loss on drying: Max. 12%