Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dow Corning® 1-2577 Conformal Coating
Dowsil™ / Dow Corning 1-2577 Conformal Coating
Dowsil™ / Dow Corning 1-2577 Conformal Coating
Dowsil™ / Dow Corning 1-2577 Conformal Coating
Catalog # 632364P
Supplier:
DOW CORNING
CAS Number:
Certificates
Expand image
Dowsil™ / Dow Corning 1-2577 Conformal Coating
Catalog # 632364P
Supplier:
DOW CORNING
CAS Number:
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
500 g
Seq:
3645
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
UN:
1993
ADR:
3,II
Product Details
Documentation