Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Cation Multi Component Standard 2 in Water with a Trace of Nitric Acid for Ion Chromatography
VWR® ARISTAR®, Cation Multi Component Standard 2 in Water with a Trace of Nitric Acid for Ion Chromatography
Catalog # 458362M
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Cation Multi Component Standard 2 in Water with a Trace of Nitric Acid for Ion Chromatography
Catalog # 458362M
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Cation multi component standard 2 in water with a trace of nitric acid ARISTAR® for ion chromatography

Specifications

About this item

Cation in dilute HNO₃.

Ca 100 ppm   Mg 100 ppm
K 100 ppm   Na 100 ppm
Li 100 ppm   NH4 100 ppm

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.