Order Entry
Austria
ContactUsLinkComponent
VWR®, Cation multi component standard 1 in water with a trace of nitric acid ARISTAR® for ion chromatography
VWR®, Cation multi component standard 1 in water with a trace of nitric acid ARISTAR® for ion chromatography
Catalog # 458352K
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Cation multi component standard 1 in water with a trace of nitric acid ARISTAR® for ion chromatography
Catalog # 458352K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Cation multi component standard 1 in water with a trace of nitric acid ARISTAR® for ion chromatography

Specifications

About this item

Cation in dilute HNO₃.

Ca 1000 ppm   Mg 200 ppm
K 200 ppm   Na 200 ppm
Li 50 ppm   NH4 400 ppm

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.