Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Calibration Standard 1 in dil. Nitric Acid for ICP-MS
VWR® ARISTAR®, Multi-Element Calibration Standard 1 in dil. Nitric Acid for ICP-MS
Catalog # 456622F
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Calibration Standard 1 in dil. Nitric Acid for ICP-MS
Catalog # 456622F
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element calibration standard 1 in dil. nitric acid ARISTAR® for ICP-MS

Specifications

About this item

17 Elements in 5% nitric acid solution.

17 Elements 10 mg/ml : Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.