VWR® ARISTAR®, Multi-Element Calibration Standard 2 in dil. Nitric Acid for ICP-MS
Catalog # 456502V
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Multi-element calibration standard 2 in dil. nitric acid ARISTAR® for ICP-MS
Specifications
About this item
29 Elements in 5% nitric acid solution.
29 Elements 10 mg/L : Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.