Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 2 in dilute Nitric Acid
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 2 in dilute Nitric Acid
Catalog # 456462H
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 2 in dilute Nitric Acid
Catalog # 456462H
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element calibration standard 2 in dil. nitric acid ARISTAR® for ICP

Specifications

About this item

In 2% nitric acid solution.

3 elements: 250 mg/L Li; 1000 mg/L Na; 10000 mg/L K

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with Certificate of Analysis.