Order Entry
Austria
ContactUsLinkComponent
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) for molecular biology
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) for molecular biology
Catalog # 438542E
Supplier:  VWR Chemicals
CAS Number:  
undefined
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) for molecular biology
Catalog # 438542E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White powder
  • Solubility in dimethylformamide (2 %):
    Clear and colourless
  • Assay:
    TLC (single spot)
  • Optical rotation:
    -92.0 to -88.0 °
  • Water:
    Max. 3.5 %
  • 1H-NMR:
    Passes test

Specification Test Results

About this item

 :
 :521.79 g/mol
Melting Pt:230 °C
Storage Temperature:Freezer
CAS Number:114162-64-0
Formula:C₂₀H₂₆BrClN₂O₇
MDL Number:MFCD00058543