Order Entry
Austria
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%
Supplier:  VWR Chemicals
Certificates
undefined
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Appearance:
    White crystalline powder
  • IR Spectrum:
    Passes test
  • Specific optical rotation (1 %; water):
    -33 to -31 °
  • Dioxane:
    Max. 1 ppm
  • Water:
    Max. 1 %

Specification Test Results

Related Information

Frequently Bought Together