VWR® TISAB II Solution (Total Ionic Strength Adjustment Buffer) for Fluoride Determinations
TISAB II solution for fluoride determinations, Plastic container, 5 L
Anbieter: VWR Chemicals
VWR® TISAB II Solution (Total Ionic Strength Adjustment Buffer) for Fluoride Determinations
Artikelnr. 160847G
Anbieter: VWR Chemicals
CAS Nummer:
Jump to:
Product Details & Documents
Total ionic strength adjustment buffer solution for use with selective ion electrodes.
Documents
Specifications
Specifications
- Pack type:Plastic container
- Pk:5 L
- Description:TISAB II solution for fluoride determinations