Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Irritant
Environment
Ammonia solution 32%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153312K
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Environment
Ammonia solution 32%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153312K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    28 - 32 %
  • Non-volatile residue:
    Max. 20 ppm
  • Fe (Iron):
    Max. 0.2 ppm
  • Pb (Lead):
    Max. 0.05 ppm
  • Transmittance (215 nm) (1 mol/l):
    Min. 10 %
  • Transmittance (254 nm) (1 mol/l):
    Min. 97 %

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650