Order Entry
Austria
ContactUsLinkComponent
Silica gel, DAVISIL® 923 suitable for use in the testing of petroleum products by IP and ASTM methods
Silica gel, DAVISIL® 923 suitable for use in the testing of petroleum products by IP and ASTM methods
Catalog # 15173LX
Supplier:  VWR Chemicals
CAS Number:  
undefined
Silica gel, DAVISIL® 923 suitable for use in the testing of petroleum products by IP and ASTM methods
Catalog # 15173LX
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Surface area:
    430 - 530 m²/g
  • pH (5 %):
    5.5 - 7.0
  • Volatile matter:
    4.5 - 10.0 %
  • Fe (as Fe2O3):
    Max. 50 ppm
  • Particle size ( 60 mesh):
    Max. 0 %
  • Particle size ( 80 mesh):
    Max. 1.2 %
  • Particle size ( 100 mesh):
    Max. 5.0 %
  • Particle size ( 200 mesh):
    Max. 15.0 %

Specification Test Results

About this item

CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
 :60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:> 1600 °C
Storage Temperature:Ambient