ContactUsLinkComponent
Nanomat 4, TLC plate applicator
Nanomat 4, TLC plate applicator
Catalog # 552-0007
Supplier:  CAMAG
undefined
Nanomat 4, TLC plate applicator
Catalog # 552-0007
Supplier:  CAMAG
Specifications
  • Description:
    Nanomat 4, without capillary pipettes and capillary holder
  • Pk:
    1
Specifications
About this item
The Nanomat 4 is used for easy application of samples in the form of spots onto TLC and HPTLC layers. A notch system ensures that sample positioning meets the requirements for automatic scanning with a densitometer. Samples are dispensed in volumes of 0,5 to 5 µl with disposable capillaries loaded into the universal capillary holder (552-0044) using the capillary guide (552-0039).
  • Easy to use and convenient
  • Fast, accurate reproducible spotting
  • Suitable for TLC and HPTLC plates