ContactUsLinkComponent
Counting chambers, Nageotte
Counting chambers, Nageotte
Catalog # 631-0928
undefined
Counting chambers, Nageotte
Catalog # 631-0928
Specifications
  • Depth of chamber:
    0,5 mm
  • Description:
    Counting chamber
  • Pk:
    1
Specifications
About this item
These counting chambers are used for special applications in lumbar fluid.
  • Subdivision: 40 rectangles of 10×0,25 mm each
  • Centre of ruling marked by double lines