Order Entry
Austria
ContactUsLinkComponent
Cation exchange chromatography columns, HiTrap SP HP
Cation exchange chromatography columns, HiTrap SP HP
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap SP HP
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 17-1152-01
  • 17-1151-01
  • 29-0513-24
  • Description:
  • HiTrap™ SP HP
  • HiTrap™ SP HP
  • HiTrap™ SP HP
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 4 - 13
  • 4 - 13
  • 4 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate

Specifications

Frequently Bought Together