Absorbent pad kits
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Handy dispenser kits allow one-handed dispensing of cellulose absorbent pads.
- One-handed operation
- Holds up to 100 pads
- Sterile pads are sterilized by gamma irradiation
- No hazardous EtO residuals
Dispense a clean cellulose pad into a petri dish without touching the pad.
Dispenser holds one tube of 100 absorbent pads. Tubes drop quickly into the hand dispenser for easy use. Absorbent pads are ideal for absorbing 2 ml of broth media to culture colonies in accordance with the membrane filter test method.
Pad Ø: 45.5 mm (1³/₄").
Pad thickness: 35 mil.
Ordering information: Each kit includes one dispenser and 10 tubes, 100 pads per tube.