VWR® shiroGEL Modular mini vertical PAGE systems
Product Details & Documents
The shiroGEL mini vertical PAGE system is modular, allowing PAGE and electroblotting to be carried out in the same tank, simply by changing the inserts. The system features inserts for each application, and a common buffer tank in which to run them. Each insert has its own electrode assembly, which connects to the lid of the buffer tank. For safety, the power leads connect directly to the lid. When the lid is removed, power is disconnected from the system. The inserts also have a small pad that fits into position in the lid. This ensures that the lid is always properly placed and aids in its removal. The buffer tank is moulded to prevent separation and leaking. A variety of spacers, combs and other accessories are available to customise the system. All spacers and combs are colour coded or clearly labelled to indicate thickness.
- Runs up to four gels simultaneously
- Gels can be cast and run using the PAGE module
- Sturdy, moulded buffer chamber with lid that fits only one way
*Multi-channel pipette compatible; **Buffer tank.
: Complete system includes buffer tank; PAGE module; casting base; 10×10 cm notched glass plates (2); 10×10 cm glass plates with bonded spacers (2); and 2×12-tooth, 1 mm thick combs.
Specifications
- Description:Mini PAGE complete system, shiroGEL
- Seq:01
- Cat. No.:700-0292
- Sample capacity:20 samples per gel, 2 gels per run
- Gel size W×H:100×100 mm
- Unit W×H×D:190×130×150 mm
- Buffer volume, gel tank:250 ml
Accessories
Product Information
- Catalog #AvailabilityPrice
- Dummy plate, 10×10 cm
Product Information
- Catalog #AvailabilityPrice
- Spacers, 2,0 mm thick
Product Information
- Catalog #AvailabilityPrice
- Spacers, 1,0 mm thick
Product Information
- Catalog #AvailabilityPrice
- Spacers, 1,5 mm thick
Product Information
- Catalog #AvailabilityPrice
- PAGE insert only (no accessories)
Product Information
- Catalog #AvailabilityPrice
- Casting stand for PAGE insert
Product Information
- Catalog #AvailabilityPrice
- Plain glass plates, 2 mm thick
Product Information
- Catalog #AvailabilityPrice
- Plain glass plates with 1,5 mm spacers, pack of 2Plain glass plates with 1,5 mm spacers, pack of 2700-0325Log in for availability1 * 1 ST - 65,60 €
Product Information
- Catalog #AvailabilityPrice
- Notched glass plates, 2 mm thick
Product Information
- Catalog #AvailabilityPrice
- Notched glass plates with 0,75 mm spacers, pack of 2Notched glass plates with 0,75 mm spacers, pack of 2700-0319Log in for availability1 * 1 ST - 71,80 €



