ContactUsLinkComponent
Vacuum Pumps and Modules for Vacuum Drying Chambers, VD and VDL
Catalog # BIND5013-0249
Supplier:  Binder
New Product
Vacuum Pumps and Modules for Vacuum Drying Chambers, VD and VDL
Catalog # BIND5013-0249
Supplier:  Binder
New Product
Specifications
  • Description:
    Vacuum pump VAP 2.1, 230 V/50 - 60 Hz option model, Chemical membrane pump with a nominal air flow of 4,0 m³/h, final pressure <2 mbar
  • Seq:
    09
  • Pk:
    1