ContactUsLinkComponent
VWR®, Cetane Number standard, ISO 5165 / EN 61715
VWR®, Cetane Number standard, ISO 5165 / EN 61715
Catalog # 99128.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Cetane Number standard, ISO 5165 / EN 61715
Catalog # 99128.290
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    Cetane Number Standard, nominal value 52,6
  • Test method:
    ISO 5165 / EN 61715
Specifications
About this item
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Cetane Number ISO 5165 / EN 61715.
  • Certified in accordance with ASTM / EN or IP test method protocols
  • Certified by Round Robin method using independent test laboratories
  • Ensures good inter-laboratory correlation
  • Tamper-evident security pack