Order Entry
Austria
ContactUsLinkComponent
VWR® Hitachi, Analyser cups for Hitachi systems
VWR® Hitachi, Analyser cups for Hitachi systems
Catalog # SIMPB720-8
Supplier:  Simport Scientific
VWR® Hitachi, Analyser cups for Hitachi systems
Catalog # SIMPB720-8
Supplier:  Simport Scientific

Specifications

  • Ext.Ø:
    12,7 mm
  • Packed:
    Bags of 1000
  • Capacity:
    2 ml
  • Description:
    Analyser cups for Hitachi systems
  • Pk:
    10.000

Specifications

About this item

These analyser cups are made of PS.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904