Orders LinkContactUsLinkComponent
Microarray molds kits, T-Sue™
Microarray molds kits, T-Sue™
Supplier:   Simport Scientific
Microarray molds kits, T-Sue™
Supplier:  Simport Scientific
Certificates
Microarray molds kits, T-Sue™
Microarray molds kits, T-Sue™
19 Options Available Below
Product Details & Documents
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to +250 °C
  • View many different samples on the same slide
  • Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed.
They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Delivery information: Each kit includes ONE T-Sue™ array mold and FOUR punch needles with stylet allowing easy removal of tissue cores for insertion.
Product Family Options
T-Sue™ Microarray molds
Product Information
  • Availability
    Price
  • SIMPM473-15
    T-Sue™ microarray mold, 15 mold cores, 4 mm core diameter
    T-Sue™ microarray mold, 15 mold cores, 4 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
  • SIMPM473-24
    T-Sue™ microarray mold, 24 mold cores, 2 mm core diameter
    T-Sue™ microarray mold, 24 mold cores, 2 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
  • SIMPM473-36
    T-Sue™ microarray mold, 36 mold cores, 3 mm core diameter
    T-Sue™ microarray mold, 36 mold cores, 3 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
  • SIMPM473-60
    T-Sue™ microarray mold, 60 mold cores, 2 mm core diameter
    T-Sue™ microarray mold, 60 mold cores, 2 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
  • SIMPM473-72
    T-Sue™ microarray mold, 72 mold cores, 1,5 mm core diameter
    T-Sue™ microarray mold, 72 mold cores, 1,5 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
  • SIMPM473-150
    T-Sue™ microarray mold, 150 mold cores, 1,5 mm core diameter
    T-Sue™ microarray mold, 150 mold cores, 1,5 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
  • SIMPM473-170
    T-Sue™ microarray mold, 170 mold cores, 1 mm core diameter
    T-Sue™ microarray mold, 170 mold cores, 1 mm core diameter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
Precast T-Sue™ paraffin blocks
Product Information
  • Availability
    Price
  • SIMPM473-15PC
    Precast T-Sue™ paraffin block, 15 mold cores, 4 mm core diameter
    Precast T-Sue™ paraffin block, 15 mold cores, 4 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-24PC
    Precast T-Sue™ paraffin block, 24 mold cores, 2 mm core diameter
    Precast T-Sue™ paraffin block, 24 mold cores, 2 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-36PC
    Precast T-Sue™ paraffin block, 36 mold cores, 3 mm core diameter
    Precast T-Sue™ paraffin block, 36 mold cores, 3 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-60PC
    Precast T-Sue™ paraffin block, 60 mold cores, 2 mm core diameter
    Precast T-Sue™ paraffin block, 60 mold cores, 2 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-72PC
    Precast T-Sue™ paraffin block, 72 mold cores, 1,5 mm core diameter
    Precast T-Sue™ paraffin block, 72 mold cores, 1,5 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-150PC
    Precast T-Sue™ paraffin block, 150 mold cores, 1,5 mm core diameter
    Precast T-Sue™ paraffin block, 150 mold cores, 1,5 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
  • SIMPM473-170PC
    Precast T-Sue™ paraffin block, 170 mold cores, 1 mm core diameter
    Precast T-Sue™ paraffin block, 170 mold cores, 1 mm core diameter
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 422,00 €
    422,00 €
    1 * 6 ST
       422,00 €
T-Sue™ punch needles
Product Information
  • Availability
    Price
  • SIMPM473-P10MA
    T-Sue™ punch needles, 1 mm diameter, magenta
    T-Sue™ punch needles, 1 mm diameter, magenta
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 166,00 €
    166,00 €
    1 * 4 ST
       166,00 €
  • SIMPM473-P15Y
    T-Sue™ punch needles, 1,5 mm diameter, yellow
    T-Sue™ punch needles, 1,5 mm diameter, yellow
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 166,00 €
    166,00 €
    1 * 4 ST
       166,00 €
  • SIMPM473-P20R
    T-Sue™ punch needles, 2 mm diameter, red
    T-Sue™ punch needles, 2 mm diameter, red
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 166,00 €
    166,00 €
    1 * 4 ST
       166,00 €
  • SIMPM473-P30P
    T-Sue™ punch needles, 3 mm diameter, pink
    T-Sue™ punch needles, 3 mm diameter, pink
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 166,00 €
    166,00 €
    1 * 4 ST
       166,00 €
  • SIMPM473-P40B
    T-Sue™ punch needles, 4 mm diameter, blue
    T-Sue™ punch needles, 4 mm diameter, blue
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 166,00 €
    166,00 €
    1 * 4 ST
       166,00 €