Order Entry
Austria
ContactUsLinkComponent
Viral RNA Xpress kit, Mag-Bind®
Viral RNA Xpress kit, Mag-Bind®
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Viral RNA Xpress kit, Mag-Bind®
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Viral RNA
  • Sample type:
    Swabs
  • Target molecule:
    RNA
  • Description:
    Mag-Bind® Viral RNA Xpress Kit

Specifications

Related Information

Frequently Bought Together