VWR®, Distillation standards for the petroleum industry, Certified Reference Materials
Supplier: VWR Chemicals
Certificates
About this item
Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for distillation at atmospheric pressure according to ASTM D86, IP 123, ISO 3405 and other such equivalent international methodology.
- Certified in accordance with ASTM / EN or IP test method protocols
- Certified by Round Robin Method using independent test laboratories ensuring good inter-laboratory correlation
- Low level of uncertainty achieves maximum data accuracy
- Fully traceable to International standards
- Supplied in tamper-evident bottles with 12 months shelf life
Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot NumberSDS_EN_CH_VWRC99106.pdf SDS_EN_CH_VWRC99107.pdf SDS_EN_CH_VWRC99108.pdf SDS_EN_DE_VWRC99106.pdf SDS_EN_DE_VWRC99107.pdf
SDS
More
VWR® for Calibrations
Product Literature
Frequently Bought Together