Orders LinkContactUsLinkComponent
Microscope objective lens
Microscope objective lens
Supplier:   LEICA MICROSYSTEMS
Microscope objective lens
Supplier:  LEICA MICROSYSTEMS
Certificates
Microscope objective lens
Microscope objective lens
55 Options Available Below
Product Details & Documents
The objective lens is a critical part of the microscope optics. The microscope objective is positioned near the sample, specimen, or object being observed. It has a very important role in imaging, as it forms the first magnified image of the sample.
  • The objective forms the first magnified image of the sample
The numerical aperture (NA) of the objective indicates its ability to gather light and largely determines the microscope’s resolution, the ability to distinguish fine details of the sample.
Objectives are categorised into performance classes on the basis of their transmission, chromatic correction, planarity, and others. From the point of view of the International Organization of Standardisation (ISO), there are three groups of objective classes differing in quality of chromatic correction (Achromats, Semi-Apochromats and Apochromats).
Product Family Options
Achromatic lens, HI PLAN
Product Information
  • Magnification
    Working distance
    Availability
    Price
  • 630-3310
    HI PLAN objective CY 10×/0,25 NA PH1; BF, PH, DF, FLUO contrast; transmitted light
    HI PLAN objective CY 10×/0,25 NA PH1; BF, PH, DF, FLUO contrast; transmitted light
    10×
    17,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 401,86 €
    401,86 €
    1 * 1 ST
       401,86 €
  • LEIM11556060
    HI PLAN 4×/0,10 NA POL -/ 121; BF, POL, FLUO contrast; transmitted light
    HI PLAN 4×/0,10 NA POL -/ 121; BF, POL, FLUO contrast; transmitted light
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 180,18 €
    180,18 €
    1 * 1 ST
       180,18 €
  • LEIM11506227
    HI PLAN 4×/0,10 NA SL ; BF, FLUO contrast; transmitted light
    HI PLAN 4×/0,10 NA SL ; BF, FLUO contrast; transmitted light
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 195,47 €
    195,47 €
    1 * 1 Pack.
       195,47 €
  • LEIM11556511
    HI PLAN 10×/0,25 NA POL disp, stain; BF, POL, DF, FLUO contrast; transmitted light
    HI PLAN 10×/0,25 NA POL disp, stain; BF, POL, DF, FLUO contrast; transmitted light
    10×
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 730,55 €
    730,55 €
    1 * 1 ST
       730,55 €
  • LEIM11556061
    HI PLAN 10×/0,25 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    HI PLAN 10×/0,25 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    10×
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 266,45 €
    266,45 €
    1 * 1 ST
       266,45 €
  • LEIM11556071
    HI PLAN 20×/0,40 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    HI PLAN 20×/0,40 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    20×
    0,92 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 348,35 €
    348,35 €
    1 * 1 ST
       348,35 €
  • LEIM11506277
    HI PLAN 20×/0,40 NA SL; BF, DF, FLUO contrast; transmitted light
    HI PLAN 20×/0,40 NA SL; BF, DF, FLUO contrast; transmitted light
    20×
    0,92 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 441,17 €
    441,17 €
    1 * 1 Pack.
       441,17 €
  • LEIM11506276
    HI PLAN 20×/0,40 NA; BF, DF, FLUO contrast; transmitted light
    HI PLAN 20×/0,40 NA; BF, DF, FLUO contrast; transmitted light
    20×
    0,92 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 197,65 €
    197,65 €
    1 * 1 ST
       197,65 €
  • LEIM11556065
    HI PLAN 40×/0,65 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    HI PLAN 40×/0,65 NA POL; BF, POL, DF, FLUO contrast; transmitted light
    40×
    0,36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 567,84 €
    567,84 €
    1 * 1 ST
       567,84 €
  • LEIM11506237
    HI PLAN 63×/0,75 NA, M25 thread; BF, DF, FLUO contrast; transmitted light
    HI PLAN 63×/0,75 NA, M25 thread; BF, DF, FLUO contrast; transmitted light
    63×
    0,31 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 411,68 €
    411,68 €
    1 * 1 ST
       411,68 €
  • LEIM11506386
    HI PLAN 100×/0,80; BF, DF, FLUO contrast; transmitted light
    HI PLAN 100×/0,80; BF, DF, FLUO contrast; transmitted light
    100×
    0,3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 562,38 €
    562,38 €
    1 * 1 ST
       562,38 €
  • LEIM11506236
    HI PLAN achromatic objective, 40×/0,65 NA; BF, DF, FLUO contrast; transmitted light
    HI PLAN achromatic objective, 40×/0,65 NA; BF, DF, FLUO contrast; transmitted light
    40×
    0,35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 250,07 €
    250,07 €
    1 * 1 ST
       250,07 €
  • LEIM11506401
    HI PLAN CY 10×/0,25 NA SL ; BF, DF, FLUO contrast; transmitted light
    HI PLAN CY 10×/0,25 NA SL ; BF, DF, FLUO contrast; transmitted light
    10×
    17,7 mm
    Log in for availability
    Log in for availability
    - 317,77 €
    317,77 €
       317,77 €
  • 630-3368
    HI PLAN EPI 5×/0,12 NA; BF, DF, FLUO contrast; transmitted, incident light
    HI PLAN EPI 5×/0,12 NA; BF, DF, FLUO contrast; transmitted, incident light
    11,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 176,90 €
    176,90 €
    1 * 1 Pack.
       176,90 €
  • 630-3366
    HI PLAN EPI 10×/0,25 NA; BF, FLUO contrast; transmitted, incident light
    HI PLAN EPI 10×/0,25 NA; BF, FLUO contrast; transmitted, incident light
    10×
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 266,45 €
    266,45 €
    1 * 1 Pack.
       266,45 €
  • 630-3367
    HI PLAN EPI 20×/0,40 NA; BF, DF, FLUO contrast; transmitted, incident light
    HI PLAN EPI 20×/0,40 NA; BF, DF, FLUO contrast; transmitted, incident light
    20×
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 348,35 €
    348,35 €
    1 * 1 Pack.
       348,35 €
  • 630-3302
    HI PLAN phase 4×/0,10 NA PH0; BF, PH, FLUO contrast; transmitted light
    HI PLAN phase 4×/0,10 NA PH0; BF, PH, FLUO contrast; transmitted light
    13,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,12 €
    215,12 €
    1 * 1 ST
       215,12 €
  • 630-3303
    HI PLAN phase 10×/0,22 NA PH1; BF, DF, PH, FLUO contrast; transmitted light
    HI PLAN phase 10×/0,22 NA PH1; BF, DF, PH, FLUO contrast; transmitted light
    10×
    7,8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,98 €
    248,98 €
    1 * 1 ST
       248,98 €
  • 630-3304
    HI PLAN phase 20×/0,30 NA PH1; BF, PH, FLUO contrast; transmitted light
    HI PLAN phase 20×/0,30 NA PH1; BF, PH, FLUO contrast; transmitted light
    20×
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 669,40 €
    669,40 €
    1 * 1 ST
       669,40 €
  • 630-3305
    HI PLAN phase 40×/0,50 NA PH1; BF, DF, PH, FLUO contrast; transmitted light
    HI PLAN phase 40×/0,50 NA PH1; BF, DF, PH, FLUO contrast; transmitted light
    40×
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 832,10 €
    832,10 €
    1 * 1 ST
       832,10 €
  • LEIM11506273
    HI PLAN phase 40×/0,50 NA PH2; BF, DF, PH, FLUO contrast; transmitted light
    HI PLAN phase 40×/0,50 NA PH2; BF, DF, PH, FLUO contrast; transmitted light
    40×
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 909,64 €
    909,64 €
    1 * 1 ST
       909,64 €
Achromatic lens, N PLAN
Product Information
  • Magnification
    Working distance
    Availability
    Price
  • LEIM11506083
    N PLAN 2,5×/0,07 NA; BF, POL, PH, FLUO contrast; transmitted, incident light
    N PLAN 2,5×/0,07 NA; BF, POL, PH, FLUO contrast; transmitted, incident light
    2,5×
    11,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 434,62 €
    434,62 €
    1 * 1 ST
       434,62 €
  • LEIM11556036
    N PLAN 2,5×/0,07 NA POL -/- 11,2; BF, POL, FLUO contrast; transmitted, incident light
    N PLAN 2,5×/0,07 NA POL -/- 11,2; BF, POL, FLUO contrast; transmitted, incident light
    2,5×
    11,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 187,00 €
    1 187,00 €
    1 * 1 ST
       1 187,00 €
  • LEIM11566077
    N PLAN EPI 5×/0,12 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast
    N PLAN EPI 5×/0,12 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast
    14 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 737,10 €
    737,10 €
    1 * 1 ST
       737,10 €
  • LEIM11506302
    N PLAN 5×/0,12 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    N PLAN 5×/0,12 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    14 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 290,47 €
    290,47 €
    1 * 1 ST
       290,47 €
  • LEIM11566083
    N PLAN EPI 10×/0,25 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast; transmitted light
    N PLAN EPI 10×/0,25 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast; transmitted light
    10×
    16,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 948,95 €
    948,95 €
    1 * 1 ST
       948,95 €
  • LEIM11506405
    N PLAN 10×/0,25 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    N PLAN 10×/0,25 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    10×
    17,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 553,64 €
    553,64 €
    1 * 1 ST
       553,64 €
  • 630-3189
    N PLAN 10×/0,25 NA PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    N PLAN 10×/0,25 NA PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    10×
    17,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 602,78 €
    602,78 €
    1 * 1 Pack.
       602,78 €
  • LEIM11566067
    N PLAN EPI 20×/0,40 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast; transmitted light
    N PLAN EPI 20×/0,40 BD; TL-BF, TL-DIC, TL-P, TL-DF, IL-BF, IL-DIC, IL-P, IL-DF, FLUO contrast; transmitted light
    20×
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 935,84 €
    935,84 €
    1 * 1 ST
       935,84 €
  • LEIM11506096
    N PLAN 20×/0,40 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    N PLAN 20×/0,40 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    20×
    0,39 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 532,90 €
    532,90 €
    1 * 1 ST
       532,90 €
  • 630-3188
    N PLAN L 20×/0,35 NA PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    N PLAN L 20×/0,35 NA PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    20×
    6,9 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 683,86 €
    1 683,86 €
    1 * 1 Pack.
       1 683,86 €
  • LEIM11506097
    N PLAN 40×/0,65 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    N PLAN 40×/0,65 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    40×
    0,36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 562,38 €
    562,38 €
    1 * 1 ST
       562,38 €
  • 630-3301
    N PLAN L phase 40×/0,55 NA PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    N PLAN L phase 40×/0,55 NA PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted light
    40×
    3,3 - 1,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 672,12 €
    2 672,12 €
    1 * 1 ST
       2 672,12 €
  • LEIM11506314
    N PLAN 40×/0,75 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    N PLAN 40×/0,75 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    40×
    0,37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 758,94 €
    758,94 €
    1 * 1 ST
       758,94 €
  • LEIM11506321
    N PLAN 50×/0,9 NA oil; BF, POL, FLUO contrast; transmitted light
    N PLAN 50×/0,9 NA oil; BF, POL, FLUO contrast; transmitted light
    50×
    0,14 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 729,73 €
    1 729,73 €
    1 * 1 ST
       1 729,73 €
  • 630-3369
    N PLAN EPI 50×/0,75 NA; BF, DIC, POL, DF, FLUO contrast; transmitted, incident light
    N PLAN EPI 50×/0,75 NA; BF, DIC, POL, DF, FLUO contrast; transmitted, incident light
    50×
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 099,64 €
    1 099,64 €
    1 * 1 Pack.
       1 099,64 €
  • LEIM11506184
    N PLAN 63×/0,80 NA 0,7/D 0,26; BF, DIC, POL, FLUO contrast; transmitted light
    N PLAN 63×/0,80 NA 0,7/D 0,26; BF, DIC, POL, FLUO contrast; transmitted light
    63×
    0,26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 959,87 €
    959,87 €
    1 * 1 ST
       959,87 €
  • LEIM11506158
    N PLAN 100×/1,25 NA oil; BF, DF, PH, FLUO contrast; transmitted light
    N PLAN 100×/1,25 NA oil; BF, DF, PH, FLUO contrast; transmitted light
    100×
    0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 017,74 €
    1 017,74 €
    1 * 1 ST
       1 017,74 €
  • 630-3370
    N PLAN EPI 100×/0,85 NA; BF, DIC, POL, DF, FLUO contrast; transmitted, incident light
    N PLAN EPI 100×/0,85 NA; BF, DIC, POL, DF, FLUO contrast; transmitted, incident light
    100×
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 1 694,78 €
    1 694,78 €
    1 * 1 Pack.
       1 694,78 €
Semi-Apochromat, PL FLUOTAR (PL-FL)
Product Information
  • Magnification
    Working distance
    Availability
    Price
  • LEIM11506413
    HC PL FLUOTAR 4×/0,13 PH0, TL-BF, TL-P, TL-DF, TL-PH, FLUO contrast; transmitted light
    HC PL FLUOTAR 4×/0,13 PH0, TL-BF, TL-P, TL-DF, TL-PH, FLUO contrast; transmitted light
    17 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 954,72 €
    954,72 €
    1 * 1 ST
       954,72 €
  • LEIM11506410
    HC PL APO 10×/0,45, TL-BF, TL-DIC, TL-DF, TL-IMC, FLUO contrast; transmitted light
    HC PL APO 10×/0,45, TL-BF, TL-DIC, TL-DF, TL-IMC, FLUO contrast; transmitted light
    10×
    2,8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 970,24 €
    2 970,24 €
    1 * 1 ST
       2 970,24 €
  • LEIM11506411
    HC PL APO 10×/0,45 PH1, TL-BF, TL-DIC, TL-DF, TL-PH, TL-IMC, FLUO contrast; transmitted light
    HC PL APO 10×/0,45 PH1, TL-BF, TL-DIC, TL-DF, TL-PH, TL-IMC, FLUO contrast; transmitted light
    10×
    2,8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 122,56 €
    4 122,56 €
    1 * 1 ST
       4 122,56 €
  • LEIM11506519
    HC PLAN FLUOTAR 20×/0,55 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    HC PLAN FLUOTAR 20×/0,55 NA; BF, DIC, POL, DF, FLUO contrast; transmitted light
    20×
    1,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 686,05 €
    1 686,05 €
    1 * 1 ST
       1 686,05 €
  • LEIM11506529
    HC PLAN APO 20×/0,80 NA, TL-BF, TL-DIC, FLUO contrast; transmitted light
    HC PLAN APO 20×/0,80 NA, TL-BF, TL-DIC, FLUO contrast; transmitted light
    20×
    0,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 495,36 €
    5 495,36 €
    1 * 1 ST
       5 495,36 €
  • LEIM11506243
    HC PLAN FL L 20×/0,40 NA CORR PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    HC PLAN FL L 20×/0,40 NA CORR PH1; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    20×
    7,5 - 6,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,16 €
    2 240,16 €
    1 * 1 ST
       2 240,16 €
  • 630-3309
    HC PLAN FL L 40×/0,60 NA CORR PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    HC PLAN FL L 40×/0,60 NA CORR PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    40×
    3,3 - 1,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 958,23 €
    2 958,23 €
    1 * 1 ST
       2 958,23 €
  • LEIM11506208
    HC PL FLUOTAR L 40×/0.60 CORR XT; TL-BF, TL-DIC, TL-P, TL-DF, TL-IMC, FLUO; Ablation, LMD, XT Light
    HC PL FLUOTAR L 40×/0.60 CORR XT; TL-BF, TL-DIC, TL-P, TL-DF, TL-IMC, FLUO; Ablation, LMD, XT Light
    40×
    3,3 - 1,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 152,24 €
    3 152,24 €
    1 * 1 ST
       3 152,24 €
  • LEIM11506382
    HC PLAN FLUOTAR 40×/0,80 NA; BF, DIC, POL, FLUO contrast; transmitted light
    HC PLAN FLUOTAR 40×/0,80 NA; BF, DIC, POL, FLUO contrast; transmitted light
    40×
    0,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 061,70 €
    2 061,70 €
    1 * 1 ST
       2 061,70 €
  • LEIM11506217
    HC PLAN FL L 63×/0,70 NA CORR PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    HC PLAN FL L 63×/0,70 NA CORR PH2; BF, DIC, POL, DF, PH, FLUO contrast; transmitted, LMD light
    63×
    2,6 - 1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 135,40 €
    4 135,40 €
    1 * 1 ST
       4 135,40 €
  • LEIM11506349
    HC PLAN APO 63×/1,40 NA - 0,60 oil; BF, DIC, FLUO contrast; transmitted light
    HC PLAN APO 63×/1,40 NA - 0,60 oil; BF, DIC, FLUO contrast; transmitted light
    63×
    0,14 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 993,99 €
    5 993,99 €
    1 * 1 ST
       5 993,99 €
  • LEIM11506385
    HC PL FLUOTAR 63×/1,30 OIL PH3, TL-BF, TL-DIC, TL-P, TL-DF, TL-PH, FLUO contrast; transmitted light
    HC PL FLUOTAR 63×/1,30 OIL PH3, TL-BF, TL-DIC, TL-P, TL-DF, TL-PH, FLUO contrast; transmitted light
    63×
    0,16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 894,62 €
    5 894,62 €
    1 * 1 ST
       5 894,62 €
  • LEIM11506220
    HCX PL APO 100×/1,40 NA - 0,70 oil; BF, DIC, DF, FLUO contrast; transmitted light
    HCX PL APO 100×/1,40 NA - 0,70 oil; BF, DIC, DF, FLUO contrast; transmitted light
    100×
    0,09 mm
    Log in for availability
    Log in for availability
    1 * 1 Pack.
    1 * 1 Pack. - 5 963,41 €
    5 963,41 €
    1 * 1 Pack.
       5 963,41 €
  • LEIM11506525
    HC PLAN FLUOTAR 100×/1,32 NA oil; BF, DIC, POL, FLUO contrast; transmitted light
    HC PLAN FLUOTAR 100×/1,32 NA oil; BF, DIC, POL, FLUO contrast; transmitted light
    100×
    0,18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 865,95 €
    4 865,95 €
    1 * 1 ST
       4 865,95 €
  • LEIM11506526
    HC PLAN FLUOTAR 100×/1,32 NA oil PH3; BF, DIC, POL, PH, FLUO contrast; transmitted light
    HC PLAN FLUOTAR 100×/1,32 NA oil PH3; BF, DIC, POL, PH, FLUO contrast; transmitted light
    100×
    0,18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 170,88 €
    5 170,88 €
    1 * 1 ST
       5 170,88 €
  • LEIM11506214
    HC PL FLUOTAR 150×/0,90,TL-BF, TL-DIC, TL-P, FLUO contrast; transmitted, LMD light
    HC PL FLUOTAR 150×/0,90,TL-BF, TL-DIC, TL-P, FLUO contrast; transmitted, LMD light
    150×
    0,25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 804,16 €
    7 804,16 €
    1 * 1 ST
       7 804,16 €