Human Recombinant IL-1RA (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
- High quality
- Low endotoxin
- Affordable
IL-1RA is secreted by immune cells, epithelial cells, hepatocytes, and adipocytes to inhibit IL-1-induced inflammation. IL-1RA therapy is used to treat auto-inflammatory diseases such as diabetes and rheumatoid arthritis.
Delivery: A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Inhibition human IL-1 alpha induced proliferation of D10,G4,1 cells, ED50 ≤50 ng/ml, bioactivity ≥2,0×10⁴ units/mg
- Gene ID:P18510
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IL-1F3|IL1RN
- UniProtKB:P18510
- Protein/peptide name:IL-1RA
- Purity:≥95%
- Molecular weight:17,3 kDa
- Sequence:MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together