Order Entry
Austria
ContactUsLinkComponent
Mouse Recombinant IL-36 gamma (Animal free) (from E. coli)
Mouse Recombinant IL-36 gamma (Animal free) (from E. coli)
Certificates
undefined
Mouse Recombinant IL-36 gamma (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q8R460
  • Reconstitution instructions:
    Sterile 10 mM sterile HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    IL-1 epsilon|IL-1F9
  • Protein/peptide name:
    IL-36 gamma
  • Purity:
    ≥95%
  • Molecular weight:
    17,5 kDa
  • Sequence:
    MGRETPDFGE VFDLDQQVWI FRNQALVTVP RSHRVTPVSV TILPCKYPES LEQDKGIAIY LGIQNPDKCL FCKEVNGHPT LLLKEEKILD LYHHPEPMKP FLFYHTRTGG TSTFESVAFP GHYIASSKTG NPIFLTSKKG EYYNINFNLD IKS
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together