About this item
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources. The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic. Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
- Gene ID:P09038
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Prostatropin|HBGF-2|FGF2
- Protein/peptide name:FGF-basic 147
- Purity:≥95%
- Molecular weight:16,5 kDa
- Sequence:MPALPEDGGS GAFPPGHFKD PKRLYCKNGG FFLRIHPDGR VDGVREKSDP HIKLQLQAEE RGVVSIKGVC ANRYLAMKED GRLLASKCVT DECFFFERLE SNNYNTYRSR KYTSWYVALK RTGQYKLGSK TGPGQKAILF LPMSAKS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 75 mM sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay