Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant NRG1-B (Animal free) (from E. coli)
 
undefined
Human Recombinant NRG1-B (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Proliferation of MCF-7 cells, ED50 ≤0,3 ng/ml, bioactivity ≥3,3×10˄6 units/mg
  • Gene ID:
    Q02297-6
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    neuregulin|NGR beta 1|HRG1 beta
  • UniProtKB:
    Q02297-6
  • Protein/peptide name:
    NRG1-beta
  • Purity:
    ≥95%
  • Molecular weight:
    7,6 kDa
  • Sequence:
    MSHLVKCAEK EKTFCVNGGE CFMVKDLSNP SRYLCKCPNE FTGDRCQNYV MASFYKHLGI EFMEAE
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together