Order Entry
Austria
Orders LinkContactUsLinkComponent
Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)

Human Recombinant FGF-Basic 154 (from E. coli)

Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)
Certificates
Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)
Human Recombinant FGF-Basic 154 (from E. coli)

5 Options Available Below

Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.

  • High quality
  • Low endotoxin
  • Affordable

Caution: This product is for research use only and is not for use in humans.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-146AF-10UG
  • SHBT100-146AF-50UG
  • SHBT100-146AF-100U
  • SHBT100-146AF-500U
  • SHBT100-146AF-1MG
  • Protein/peptide name:
  • FGF-Basic 154 (AF)
  • FGF-Basic 154 (AF)
  • FGF-Basic 154 (AF)
  • FGF-Basic 154 (AF)
  • FGF-Basic 154 (AF)
  • Protein synonyms:
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P09038
  • P09038
  • P09038
  • P09038
  • P09038
  • Sequence:
  • MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • Molecular weight:
  • 17,3 kDa
  • 17,3 kDa
  • 17,3 kDa
  • 17,3 kDa
  • 17,3 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 82,30 €
    82,30 €
    1 * 10 µG
       82,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-146AF-10UG
    Protein/peptide name
    FGF-Basic 154 (AF)
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09038
    Sequence
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
    Molecular weight
    17,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 209,00 €
    209,00 €
    1 * 50 µG
       209,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-146AF-50UG
    Protein/peptide name
    FGF-Basic 154 (AF)
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09038
    Sequence
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
    Molecular weight
    17,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 298,00 €
    298,00 €
    1 * 100 µG
       298,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-146AF-100U
    Protein/peptide name
    FGF-Basic 154 (AF)
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09038
    Sequence
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
    Molecular weight
    17,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 640,00 €
    640,00 €
    1 * 500 µG
       640,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-146AF-500U
    Protein/peptide name
    FGF-Basic 154 (AF)
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09038
    Sequence
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
    Molecular weight
    17,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 1 080,00 €
    1 080,00 €
    1 * 1 mg
       1 080,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-146AF-1MG
    Protein/peptide name
    FGF-Basic 154 (AF)
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09038
    Sequence
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
    Molecular weight
    17,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.