Order Entry
Austria
ContactUsLinkComponent
Water, SupraSolv® for GC-MS, Supelco®
Water, SupraSolv® for GC-MS, Supelco®
Catalog # 1.03702.2500
Supplier:  Merck
CAS Number:  
Water, SupraSolv® for GC-MS, Supelco®
Catalog # 1.03702.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Identity (IR-spectrum):
    passes test
  • Colour:
    ≤ 10 Hazen
  • Evaporation residue:
    ≤ 5.0 mg/l
  • TOC:
    ≤ 30 ppb

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332