Order Entry
Austria
ContactUsLinkComponent
Diethyl ether stabilised, EMPARTA® ACS for analysis
Flammable
Irritant
Diethyl ether stabilised, EMPARTA® ACS for analysis
Catalog # 1.07062.4000
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether stabilised, EMPARTA® ACS for analysis
Catalog # 1.07062.4000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
 :74.12 g/mol
ADR:3,I
 :M-Clarity™ quality level = MQ200
 :Stabilised with ethanol
UN:1155
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646