About this item
Eppendorf® Conical Tubes with SnapTec® caps offer secure sample containment for diverse applications. The innovative SnapTec® Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in the lab.
- SnapTec® caps allow effortless, secure sealing for sample integrity in molecular biology and genomics
- Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation
- Well-suited for various lab tasks, ensuring adaptability across experiments
- Durable construction crafted for longevity, supporting applications like cell culture, protein purification, and more
- Streamlines processes, benefitting applications such as qPCR, cloning, and sample storage
- Available in 25 ml and SnapTec® 50 to 45 ml formats
Eppendorf® Conical Tubes with SnapTec® Caps redefine sample containment, catering to diverse lab applications. The innovative SnapTec® Cap ensures secure sealing for molecular biology and genomics, preserving sample integrity during storage. Versatile across lab tasks, they adapt to cell culture and protein purification. Crafted for durability, these tubes support longevity in various applications. Elevate scientific processes with Eppendorf®, offering trusted quality for versatile lab applications.
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Material: PP
Maximum RCF: 17000×g
Specifications
- Catalog No:
- 525-1604
- 525-1608
- 525-1609
- 525-1605
- 525-1610
- 525-1606
- 525-1607
- 525-1603
- Autoclavable:
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- When open, 121 °C, 20 min
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Dimensions:
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Ø 30 mm
- Operating temperature range:
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- −86...+100 °C
- Environmental certification:
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label