Orders LinkContactUsLinkComponent
CytoFLEX platform flow cytometers
CytoFLEX platform flow cytometers
CytoFLEX platform flow cytometers
CytoFLEX platform flow cytometers
The CytoFLEX platform flow cytometer is a revolutionary system presenting optimal excitation and emission, minimising light loss and maximising sensitivity.
  • Exquisite sensitivity (80 nm) enabling nano particle analysis including extracellular vesicles, viruses and more
  • Extensive set of repositionable band pass filters
  • Flexibility to upgrade by adding additional parameters
  • Intuitive software to facilitate multicolour analysis
The CytoFLEX platform is divided into several versions across three models. An instrument is defined by the lasers included. Each instrument is offered in a number of configurations, determined by the number of activated channels from each laser. Choose the configuration and number of parameters needed immediately, and activate additional parameters as the needs of the laboratory change in the future. Each instrument includes the full complement of repositionable band pass filters. Move filters to the activated channels based upon the wavelengths to be detected. Additional non standard band pass filters are available for even more assay flexibility.
CytoFLEX models bring up to three lasers instrument providing up to 13 fluorescent channels to accommodate most basic flow cytometry assay needs.
- Violet-Blue-Red (VB-R) Series, fully activated as few as one laser and four colours, activate additional colours and upgrade later, up to three lasers and 13 colours
- Includes 15 band pass filters
CytoFLEX S models bring up to four laser instruments to the research community expanding the fluorochrome palette for special applications. Four different series are available.
- The four series are: Violet-Blue-Yellow Green-Red (V-B-Y-R), Near UV-Violet-Blue-Red (N-V-BR), Near UV-Violet-Blue-Yellow Green (N-V-B-Y), Violet-Blue-Red-IR (V-BR-I)
- Activate as few as two lasers and four colours and upgrade later within the series, up to four lasers and 13 colours
- Includes 15 band pass filters depending on the chosen series
CytoFLEX LX models bring up to six laser instruments to the research community providing up to 21 fluorescent channels. Two different series are available, depending on choice of UV or near UV laser.
- Two series are available depending on your choice of UV or near-UV: UV-Violet-Blue-Yellow Green-Red-IR (UV-B-Y-R-I) and Near UV-Violet-Blue-Yellow Green-Red-IR (NV-B-Y-R-I)
- Activate as few as four lasers and 14 colour parameters or fully activate all six lasers for 21 colour parameters
- Includes 23 band pass filters depending on the chosen series
Additional specifications:
- Nominal acquisition rate: 30000 events per second with all configured parameters
- Signal processing: Fully digital system with seven decade data display
- Sample flow rates: Fixed flow rates: 10, 30 and 60 μl/min and custom flow rate Ccntrol mode from 10 to 240 μl/min in 1 μl increments
- Absolute counting: Gravimetric calibration for absolute counts within CytExpert software
- Software: Three different installation modes are available depending on the level of security required: Default Mode with no user login, User Management Mode and - Electronic Records Management Mode, which provides tools that facilitate compliance with 21 CFR Part 11
- Automation: An API (Application Programming Interface) is available for automation integration
- Scatter performance is optimised for resolving human lymphocytes, monocytes and granulocytes as well as nanoparticles
- Optional plate loader available to support 96-well flat and deep well plates (45,5 mm height max, 5,8 mm well bottom diameter minimum)
  • Catalog No:
  • BECMB53010
  • BECMB53011
  • BECMB53012
  • BECMB53016
  • BECMB53000
  • BECMB53001
  • BECMB53002
  • BECMB53003
  • BECMB53004
  • BECMB53005
  • BECMB53006
  • BECMB53007
  • BECMB53008
  • BECMB53009
  • BECMB53013
  • BECMB53014
  • BECMB53018
  • BECMB53037
  • BECMB96622
  • BECMC02944
  • BECMC02945
  • BECMC02946
  • BECMB75408
  • BECMB75811
  • BECMB75812
  • BECMB78557
  • BECMB78558
  • BECMB78559
  • BECMB78560
  • BECMB78561
  • BECMB96618
  • BECMB96619
  • BECMB96620
  • BECMB96621
  • BECMC02948
  • BECMC02949
  • BECMC01158
  • BECMC01159
  • BECMC01160
  • BECMC01161
  • BECMC02947
  • BECMC00445
  • BECMC00446
  • BECMC11183
  • BECMC11184
  • BECMC11185
  • BECMC11186
  • Model:
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX/CytoFLEX S
  • CytoFLEX LX
  • CytoFLEX LX
  • CytoFLEX LX
  • CytoFLEX LX
  • CytoFLEX LX
  • CytoFLEX LX
  • Sensitivity:
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
  • Resolution:
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
  • Operating temperature:
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • 15...27 °C
  • Voltage:
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • W×D×H:
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 425×425×340 mm
  • 605×733×451 mm
  • 605×733×451 mm
  • 605×733×451 mm
  • 605×733×451 mm
  • 605×733×451 mm
  • 605×733×451 mm
  • Weight:
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 23,4 kg
  • 79,0 kg
  • 79,0 kg
  • 79,0 kg
  • 79,0 kg
  • 79,0 kg
  • 79,0 kg
Product Family Options
CytoFLEX
  • Description
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53010
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53011
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53012
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53016
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 5 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53000
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53001
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 5 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53002
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53003
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 5 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53004
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53005
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 3 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53006
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 3 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53007
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53008
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53009
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53013
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 3 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53014
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53018
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 5 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB53037
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB96622
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02944
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02945
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02946
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
CytoFLEX S
  • Description
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 405 nm laser, 4 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB75408
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB75811
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB75812
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 3 fluorescence detectors 405 nm laser, 2 fluorescence detectors 375 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB78557
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 2 fluorescence detectors 375 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB78558
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 5 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 375 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB78559
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 405 nm laser, 4 fluorescence detectors 561 nm laser, 2 fluorescence detectors 375 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB78560
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 561 nm laser, 2 fluorescence detectors 375 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB78561
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB96618
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 561 nm laser, 4 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB96619
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 4 fluorescence detectors 405 nm laser, 4 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB96620
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 2 fluorescence detectors 638 nm laser, 2 fluorescence detectors 405 nm laser, 3 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMB96621
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 405 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02948
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 2 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02949
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 4 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC01158
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC01159
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 4 fluorescence detectors 405 nm laser, 2 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC01160
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 4 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser, 4 fluorescence detectors 405 nm laser, 2 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC01161
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
  • Flow cytometer, 2 fluorescence detectors 488 nm laser, 2 fluorescence detectors 405 nm laser, 2 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC02947
    Model
    CytoFLEX/CytoFLEX S
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    425×425×340 mm
    Weight
    23,4 kg
CytoFLEX LX
  • Description
  • Flow cytometer, 3 fluorescence detectors 375 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser, 5 fluorescence detectors 561 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC00445
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
  • Flow cytometer, 3 fluorescence detectors 375 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser, 5 fluorescence detectors 561 nm laser, 3 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC00446
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
  • Flow cytometer, 3 fluorescence detectors 355 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser, 3 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC11183
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
  • Flow cytometer, 3 fluorescence detectors 355 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser ,5 fluorescence detectors 561 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC11184
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
  • Flow cytometer, 3 fluorescence detectors 355 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser ,5 fluorescence detectors 561 nm laser, 3 fluorescence detectors 638 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC11185
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
  • Flow cytometer, 3 fluorescence detectors 355 nm laser, 5 fluorescence detectors 405 nm laser, 3 fluorescence detectors 488 nm laser ,5 fluorescence detectors 561 nm laser, 3 fluorescence detectors 638 nm laser, 2 fluorescence detectors 808 nm laser
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    BECMC11186
    Model
    CytoFLEX LX
    Sensitivity
    FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser
    Resolution
    488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles
    Operating temperature
    15...27 °C
    Voltage
    100 - 240 V
    W×D×H
    605×733×451 mm
    Weight
    79,0 kg
Accessories
  •  
    Description
  • BECMA04-1-0048
    Peristaltic sample tubing replacement kit
    1 * 1 KIT - 168,00 €
  •  
    Description
  • BECMB63215
    CytoFLEX plate loader
    1 * 1 ST - 22 340,00 €
  •  
    Description
  • BECMA64669
    Flow clean, 500 ml, cleaning solution for CytoFLEX
    1 * 500 mL
  •  
    Description
  • BECM81911
    Contrad 70 cleaning agent, 1×1 liter
    1 * 1 L
  •  
    Description
  • BECMA04-1-0041
    Sheath filter
    1 * 1 ST - 713,00 €
  •  
    Description
  • BECMB51503
    Sheath fluid, 10 L cubitainer
    1 * 10 L - 36,60 €
  •  
    Description
  • BECMB84161
    CytoFLEX Upgrade from V0-B3-R1 to V2-B3-R2
    1 * 1 ST
  •  
    Description
  • BECMA70449
    FlowClean cleaning agent, 500 ml
    1 * 1 ST