Avantor® ACE® Excel® Oligo, HPLC/UHPLC Columns, Analytical
Supplier: Avantor
Certificates
About this item
Avantor® ACE® Oligo is a unique encapsulated stationary phase for high resolution ion-pair reversed-phase analysis of oligonucleotides. These high pH stable stainless steel columns are compatible with all buffers and mobile phases typically used for oligonucletide analysis by LC-UV and LC-MS, including TEAA and HFIP.
- High efficiency 1,7 and 3,0 µm particle sizes for high resolution oligonucleotide separations
- Ultra-inert, base deactivated silica for exceptional reproducibility and peak shape
- Excellent column lifetime under oligonucleotide testing conditions
- Ultra-low bleed for excellent LC/MS compatibility
Specifications
- Catalog No:
- EXL-1115-1502
- EXL-1115-1002
- EXL-1115-0502
- EXL-1P15-GD4U
- EXL-1P15-GD2U
- EXL-1715-1002
- EXL-1715-0502
- EXL-1115-1546
- EXL-1115-1046
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Application:
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Carbon load:
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH range:
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Related Information
Frequently Bought Together