Order Entry
Austria
ContactUsLinkComponent
Easy DNA/RNA extraction kit, genesig®
Easy DNA/RNA extraction kit, genesig®
Catalog # GENESIGEASY-EK
Supplier:  PRIMERDESIGN
CAS Number:  
Easy DNA/RNA extraction kit, genesig®
Catalog # GENESIGEASY-EK
Supplier:  PRIMERDESIGN
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    Stool
    Swabs
    Bacteria
    Blood
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
    Food samples
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    Easy DNA/RNA Extraction Kit for 50 extractions
  • Description:
    Easy DNA/RNA Extraction Kit

Specifications

About this item

The genesig easy DNA/RNA extraction protocol begins with a simple lysis step where cells and tissue are lysed to release their nucleic acid. Then minute magnetic particles are added to bind to DNA/RNA. When placed on to the genesig magnetic separator the particles are pulled to the side of the tube making it easy to remove the unwanted supernatant with a pipette. Then a series of simple wash steps are performed before the DNA/RNA is washed off the beads back in to solution, ready for analysis by real-time PCR.

It is fast, and incredibly easy to perform.

  • Extracts DNA and RNA with high yields in 45-60 mins
  • Works with huge range of sample types
  • Safe protocol with no phenolic chemicals
  • No centrifuge required - for use in the lab or in the field

Suitable sample types:
Clinical and Veterinary - including whole blood, plasma, and serum; saliva and sputum; faeces and urine; tissues; dry swabs; and bacterial culture broth
Food - including meat, fish, milk, and cooked or processed food products
Environmental - including plant, soil, and water